Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubocurarine Chloride Pentahydrate 98.0+%, TCI America™
SDP

Catalog No. C04331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C04331G Supplier TCI America Supplier No. C04331G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6989-98-6
Molecular Formula C37H52Cl2N2O11
Molecular Weight (g/mol) 771.73
MDL Number MFCD00150157
InChI Key WMIZITXEJNQAQK-GGDSLZADSA-N
Synonym Curare
PubChem CID 16051918
IUPAC Name hydrogen (1S,16R)-9,21-dihydroxy-10,25-dimethoxy-15,15,30-trimethyl-7,23-dioxa-15,30-diazaheptacyclo[22.6.2.2³,⁶.1⁸,¹².1¹⁸,²².0²⁷,³¹.0¹⁶,³⁴]hexatriaconta-3,5,8,10,12(34),18,20,22(33),24(32),25,27(31),35-dodecaen-15-ium pentahydrate dichloride
SMILES [H+].O.O.O.O.O.[Cl-].[Cl-].COC1=CC2=C3[C@@H](CC4=CC=C(O)C(OC5=CC6=C(CCN(C)[C@H]6CC6=CC=C(OC3=C1O)C=C6)C=C5OC)=C4)[N+](C)(C)CC2

Specifications

Melting Point 268°C
Quantity 1 g
UN Number 1544
Formula Weight 681.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tubocurarine Chloride Pentahydrate
RTECSNumber YO5100000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.