Learn More
MP Biomedicals™ Tubulin, Porcine, 99% Pure
Catalog No. 08771151 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
08-771-151 1 mg
1 options
Catalog No. 08-771-151 Supplier MP Biomedicals™ Supplier No. 08771151
Add to Cart
Add to Cart
Tubulin, Porcine, 99% Pure is composed of a heterodimer of two closely related 55 kDa proteins called alpha and beta tubulin.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.