Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tubulin Tracker™ Deep Red

Catalog No. T34076
Encompass_Preferred
Change view
Click to view available options
Quantity:
300 slides
60 slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34076 300 slides
T34077 60 slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34076 Supplier Invitrogen™ Supplier No. T34076
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tubulin Tracker™ Deep Red provides deep-red/far-red fluorescence when bound to polymerized tubulin in live cells.

  • Based on Docetaxel conjugated with a bright, photostable deep-red fluorophore
  • Docetaxel belongs to the family of cytoskeletal drugs that target tubulin
  • Absorbs and emits optimally at 652 nm and 669 nm, respectively and can be visualized with standard Cy5 filter settings using almost any fluorescent imaging instrument
  • Can be multiplexed with blue, green, orange, red, and near-IR fluorophores.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains sufficient material for at least 300 slides at 1 mL per slide.

  • 5 vials Tubulin Tracker Deep Red (Add 60 μL/vial of anhydrous DMSO to make a 1000X (1mM) stock solution.  Dissolve stock solution 1:1000 in cell imaging solution to make 1X (1 μM) staining solution.)
    • 5 x 77 mg Probenecid (Dissolve vial contents in 1 mL of live cell compatible buffer to make a 100X stock solution.)

      Store at ≤–20°C after recieving.
      Desiccate and protect from light.
      When stored as directed, product is stable for at least 6 months.

Excitation Wavelength Range 652⁄669
Quantity 300 slides
Product Line TubulinTracker
Shipping Condition Approved for shipment on Wet or Dry Ice
Label Type UV-Vis Labels
Product Type Tubulin Tracker
Sub Cellular Localization Tubulin
With Tubulin Tracker Deep Red and Tubulin Tracker Green, can I fix samples after labeling?

No. The labeling is not retained upon fixation. Imaging of the samples should be done only on live cells. To limit photobleaching while viewing live cells, use ProLong Live Antifade Reagent, for live cell imaging (Cat No. P36974 and P36975).

How is Docetaxel in Tubulin Tracker Deep Red different from Taxol in Tubulin Tracker Green? Are there any differences in the binding or cytotoxicity between the two reagents?

Docetaxel has a slightly higher binding affinity than Taxol and exhibits longer retention time due to better uptake and slower efflux, but for use in labeling cultured cells, these differences should be minimal. More information comparing the properties of these two reagents is provided in this publication: https://www.ncbi.nlm.nih.gov/pubmed/1671606 https://www.cell.com/fulltext/S1074-5521(05)00301-7

Is Tubulin Tracker Deep Red acetylated? Does it require intracellular esterase activity to make the reagent active?

Tubulin Tracker Deep Red is not acetylated. It is inherently cell permeable and is not modified further by any cellular enzymatic activity to fluoresce or bind to actin.

Do I need to use Probenecid when using Tubulin Tracker Deep Red?

The use of Probenecid is optional. Its requirement is dependent upon the cell type. Some cell types actively efflux the reagent out of the cells within a short time, limiting detection, other cell types do not exhibit this activity or exhibit very limited activity.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.