Learn More
Selleck Chemical LLC Tucatinib
SDP
Supplier:  Selleck Chemical LLC S83621G
Encompass_Preferred
Tucatinib (Irbinitinib ONT-380 ARRY-380) is an oral potent selective reversible and ATP-competitive small-molecule inhibitor of ErbB-2 (also called HER2) with IC50s of 8 nM and 7 nM for ErbB-2 and p95 HER2 respectively in cell-based assays showing 500-fold selective for HER2 vs EGFR It has potential antineoplastic activity
Catalog No. 50-000-00843
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.