Learn More
Dynalon™ Tuff Vials
SCI_ED
Catalog No. S37270 Shop All Dynalab Corporation Products
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S37270 5 mL
S37271 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S37270 Supplier Dynalon™ Supplier No. 6264340005
Add to Cart
Add to Cart
Ideal for leakproof shipping of reagents and buffer solutions, for packaging of serum, veterinary, pharmaceuticals, cosmeceutical and nutraceutical extracts. Dynalon™ Tuff Vials are used with most aromatic compounds and essential oils.
  • Vials are made of break resistant polycarbonate
  • Includes polypropylene leakproof closures
  • Reusable and autoclavable
  • Clear glass helps viewing of valuable contents, liquid or solid
  • 180mL can also be used as an outer protective container
Specifications
Material Polycarbonate
Volume (Metric) 5 mL
Style Round
Closure Material Polypropylene
Closure Size 20-415