Learn More
Tufpak Secura'T™ Autoclavable Biohazard Bags
Description
- 275°F Temperature Resistance
- With Temperature Indicator Patch ("Contents Have Been Autoclaved")
- Wire Ties Included
- Exceed ASTM 1922 Tear Resistance test of 480 grams in both directions
- Exceed ASTM 1709 Dart Impact Strength test of 165 grams
- Proper Handling Instructions written in 4 languages English, French, German and Spanish
- Material: Polypropylene
- Color: Red
- Thickness: 2 mil
Compliant with:
- DOT Regulation 49CFR.197
- OSHA Regulation 1910.1030
- California Health and Safety Code 117630, Effective January 1st 2015
- Florida Dept. of Health Code # 64E-16.004-2C, Registration # PP-2-R1-04
- Texas Environmental, 30 Texas Administrative Code §330.1207(c)
- CDC Procedural Guidance on proper packaging of highly infectious waste
- EU Hazardous Waste Directive, and World Health Organization directive for Safe Management of Healthcare Waste
Specifications
Specifications
| Color | Red |
| Dimensions (L x W) | 24 x 30 in. (61 x 76 cm) |
| Autoclavable | Autoclavable |
| For Use With (Application) | Biohazard Waste bag as required by law for the safe handling of infectious materials. To render Biohazardous and Hazardous Waste to a nonInfectious state after autoclaving for safe economical disposal. |
| Thickness | 2 mil |
| Material | Polypropylene |
| Temperature (English) | 275°F |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.