Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tufpak Secura'T™ Autoclavable Biohazard Bags

Catalog No. 14375197
Change view
Click to view available options
Dimensions (L x W):
12 x 24 in. (30.5 x 61 cm)
14 x 19 in. (35.5 x 48 cm)
19 x 24 in. (48 x 61 cm)
24 x 30 in. (61 x 76 cm)
24 x 36 in.(61x 91 cm)
25 x 35 in. (63.5 x 89 cm)
31 x 38 in. (78.5 x 96.5 cm)
37 x 48 in. (94 x 122 cm)
8 x 12 in. (20 x 30.5 cm)
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
14-375-197 25 x 35 in. (63.5 x 89 cm)
14-375-192 8 x 12 in. (20 x 30.5 cm)
14-375-193 12 x 24 in. (30.5 x 61 cm)
14-375-194 14 x 19 in. (35.5 x 48 cm)
14-375-195 19 x 24 in. (48 x 61 cm)
14-375-196 24 x 30 in. (61 x 76 cm)
14-375-202 24 x 36 in.(61x 91 cm)
14-375-198 31 x 38 in. (78.5 x 96.5 cm)
14-375-199 37 x 48 in. (94 x 122 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14-375-197 Supplier Tufpak Supplier No. 12122535

These Autoclavable Biohazard Bags meet all Federal and State Regulations and Requirements for hazardous waste.

Designed to neutralize hazardous and biohazardous waste in an autoclave chamber. Rendering waste after autoclaving to a noninfectious state and more affordable disposal as defined by the Federal EPA.
  • 275°F Temperature Resistance
  • With Temperature Indicator Patch ("Contents Have Been Autoclaved")
  • Wire Ties Included
  • Exceed ASTM 1922 Tear Resistance test of 480 grams in both directions
  • Exceed ASTM 1709 Dart Impact Strength test of 165 grams
  • Proper Handling Instructions written in 4 languages English, French, German and Spanish
  • Material: Polypropylene
  • Color: Red
  • Thickness: 2 mil

 

Compliant with:

  • DOT Regulation 49CFR.197
  • OSHA Regulation 1910.1030
  • California Health and Safety Code 117630, Effective January 1st 2015
  • Florida Dept. of Health Code # 64E-16.004-2C, Registration # PP-2-R1-04
  • Texas Environmental, 30 Texas Administrative Code §330.1207(c)
  • CDC Procedural Guidance on proper packaging of highly infectious waste
  • EU Hazardous Waste Directive, and World Health Organization directive for Safe Management of Healthcare Waste

 

 

Specifications

Color Red
Dimensions (L x W) 25 x 35 in. (63.5 x 89 cm)
Autoclavable Autoclavable
For Use With (Application) Biohazard Waste bag as required by law for the safe handling of infectious materials. To render Biohazardous and Hazardous Waste to a nonInfectious state after autoclaving for safe economical disposal.
Thickness 2 mil
Material Polypropylene
Temperature (English) 275°F
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.