Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tulobuterol Hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAJ61448MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61448MD 250 mg
AAJ6144803 1 g
AAJ6144806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61448MD Supplier Thermo Scientific Chemicals Supplier No. J61448MD
Get 15% Off Thermo Scientific Reagents
Use promo code REAGENTS15
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

beta-2-adrenergic receptor agonist | CAS: 56776-01-3 | C12H18ClNO·HCl

beta-2-adrenergic receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56776-01-3
Molecular Formula C12H18ClNO·HCl

Specifications

Quantity 250 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) beta-2-adrenergic receptor agonist
Formula Weight 264.19
Sensitivity Hygroscopic
Melting Point 159°C to 163°C
Solubility Soluble in methanol or water
Product Type Signal Transduction Reagent
Form Powder
Assay 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.