Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TULP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501732295 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2295 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2295 Supplier Proteintech Group Inc Supplier No. 189711AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

TULP1 is a member of the tubby-like gene family (TULPs). Members of this family have been identified in plants, vertebrates, and invertebrates. TULP proteins share a conserved C-terminal region of approximately 200 amino acid residues. The protein encoded by this gene is thought to play a role in the physiology of photoreceptors. Mutations in this gene are associated with recessive juvenile retinitis pigmentosa and Leber congenital amaurosis-15.

Specifications

Antigen TULP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TULP1
Gene Accession No. O00294, Q9Z273
Gene Alias RP14, tubby like protein 1, Tubby related protein 1, TUBL1, TULP1
Gene Symbols Tulp1
Host Species Rabbit
Immunogen TULP1 Fusion Protein Ag5598
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22157, 309900, 7287
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.