Learn More
Description
- Tumor-derived Exosome ELISA Kit is double sandwich enzyme-linked immunoassay for tumor-derived Exosomes enrichment and quantification from biological fluids or cell media.
Specifications
Specifications
| Cross Reactivity | Human |
| Product Type | Kit |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C. Do not freeze. |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.