Learn More
Novus Biologicals Tumor-Derived Exosome Isolation Beads

Specifications
Specifications
| Description | Tumor-Derived Exosome Isolation Beads |
| Synonym | EV isolation, exosome isolation, extracellular vesicle isolation |
| Content And Storage | Store at 4°C |
| Target Species | Human |
| Quantity | 10 Reactions |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.