Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Tumor Growth Antibody Pack
SDP

Catalog No. NBP178956 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178956 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178956 Supplier R&D Systems™ Supplier No. NBP178956

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. For Research applications

Specifications

Antigen Tumor Growth
Applications Immunohistochemistry, Immunoassay
Conjugate Unconjugated
Dilution Immunohistochemistry, Knockout Validated
Formulation See individual datasheets.
Immunogen See individual datasheets.
Quantity 2 Vials
Research Discipline Cancer
Test Specificity This Tumor Growth Antibodies Sample Pack includes antibodies to YAP1 and AGR2.
Includes 1.NB110-58358: YAP1 Antibody
2.NBP1-05933: AG-2 Antibody
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.