Learn More
R&D Systems™ TumorTACS In Situ Apoptosis Detection Kit
Description
Includes
Streptavidin-HRP, DAB, Methyl Green counterstain, and reagents necessary for generating positive controls
A common biochemical marker of apoptosis is DNA fragmentation. In situ, the 3 ft. ends of cleaved DNA fragments provide a substrate for the enzyme terminal deoxynucleotidyl transferase (TdT) which adds nucleotides at the site of DNA breaks.
Optimized for detection of apoptosis in tumor-derived sections and cells
Supplemented with a cation that specifically enhances the signal in apoptotic tumor cells
TACS-Nuclease™ allows positive controls to be
generated for each experim
Specifications
Specifications
| Quantity | 30 Tests |
| Content And Storage | The product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Do not use past expiration date. |