Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten wire, gold plated, 0.05mm (0.002in) dia, W 99.95% (metals basis), Au 99.99%

Catalog No. AA41039G5
Change view
Click to view available options
Quantity:
1 m
5 m
10 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41039G5 5 m
AA41039G1 1 m
AA41039G6 10 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41039G5 Supplier Thermo Scientific Chemicals Supplier No. 041039G5
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tungsten wire, gold plated
Name Note 0.05mm (0.002 in.) dia.
Quantity 5 m
Weight ≈0.02g/m (W only), 3 to 5 wt% Au plating
Form Wire
Assay Percent Notes (metals basis)
MDL Number MFCD00011461
Percent Purity 99.95%
Concentration or Composition (by Analyte or Components) Au 99.99%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.