Learn More
Lkt Laboratories CAS 11089-65-9, ≥98%, GlcNAc phosphotransferase and glycoprotein synthesis inhibitor. It inhibits TLR activation-induced pro-inflammatory cytokine release in macrophages.
SDP
Supplier:  Lkt Laboratories T8153
Encompass_Preferred
Tunicamycin is a nucleoside antibiotic mixture that inhibits glycoprotein synthesis. Tunicamycin prevents N-glycosylation by inhibiting GlcNAc phosphotransferase and other enzymes, inducing G1 phase cell cycle arrest in vitro. Tunicamycin displays anti-inflammatory activity, inhibiting toll-like receptor (TLR) activation-induced inflammation by decreasing activation of NF-κB, activity of CRE, and expression of iNOS, COX-2, TNF-α, and IL-1β in macrophages.
Catalog No. NC2539820
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.