Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Tunicamycin, Calbiochem™,

Catalog No. 65438050MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
65-438-050MG 50 mg Glass Bottle
65-438-010MG 10 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65-438-050MG Supplier MilliporeSigma™ Supplier No. 65438050MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 11089-65-9
Molecular Formula C39H64N4O16

Specifications

Color White
Packaging Glass Bottle
Quantity 50 mg
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.