CAS RN 11089-65-9
IUPAC Name:
(2E)-N-[(2R,3R,4R,5R,6R)-6-{2-[(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl}-2-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-dihydroxyoxan-3-yl]-5-methylhex-2-enamide
Synonyms:
(2E)-N-[(2R,3R,4R,5R,6R)-6-{2-[(2R,3S,4R,5R)-5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl}-2-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-dihydroxyoxan-3-yl]-5-methylhex-2-enamide
Molecular Weight (g/mol):
718.71
Molecular Formula:
C30H46N4O16
InChi Key:
ZHSGGJXRNHWHRS-PEALBESXSA-N
SMILES:
CC(C)C\C=C\C(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CC(O)[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=CC(=O)NC2=O)O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O
1
–
6
of
6
results
1
Tunicamycin (Antibiotic), From Streptomyces species
CAS: 11089-65-9
2
Tunicamycin, MP Biomedicals™
CAS: 11089-65-9
3
Tunicamycin, 10 mg/ml in DMSO, sterile-filtered
CAS: 11089-65-9 MDL Number: MFCD00065709
4
Tunicamycin, 95%
CAS: 11089-65-9 Molecular Formula: C39H64N4O16 MDL Number: MFCD00065709
5
R&D Systems™ Tunicamycin
CAS: 11089-65-9
6
MilliporeSigma™ Tunicamycin, Calbiochem™,
CAS: 11089-65-9 Molecular Formula: C39H64N4O16