Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tunicamycin, MP Biomedicals™

Catalog No. ICN15002801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15002801 1 mg
ICN15002810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15002801 Supplier MP Biomedicals, Inc Supplier No. 0215002801
Add to Cart
Add to Cart

Chemical Identifiers

CAS 11089-65-9

Specifications

Type Tunicamycin
Physical Form Powder
Color Tan to White
Quantity 1 mg
Formula Weight 840
Chemical Name or Material Tunicamycin
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.