Learn More
Abcam Tunicamycin, Protein Glycosylation Inhibitor, Solid, 50 mg
Description
- Molecular Weight: 816.9 Da
- Purity: >98%
- Form: Solid
- Mechanism of Action: Nucleoside antibiotic that inhibits protein glycosylation by blocking GlcNAc phosphotransferase (GPT) and the transfer of N-acetylglucosamine-1-phosphate from UDP-N-acetylglucosamine to dolichol phosphate.
- Biological Activity: Inhibits N-linked glycosylation and blocks the formation of N-glycosidic protein-carbohydrate linkages; active in vitro against Gram-positive bacteria, yeasts, fungi and viruses.
- Composition: 98% Tunicamycin from Streptomyces sp.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | GlcNAc Phosphotransferase |
| Reactivity | Gram-positive bacteria, yeasts, fungi, viruses |
| Shipping Condition | Ambient |
| Form | Solid |
| Content And Storage | Store at 4°C. |
| Formulation | 98% Tunicamycin from Streptomyces sp |
| Quantity | 50 mg |
| For Use With (Application) | Protein glycosylation inhibition, N-linked glycosylation inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.