Learn More
United Scientific™ Tuning Fork Set of 13 with Storage Box
SDPSCI_ED
Catalog No. S111126 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type: Tuning Fork Set
Catalog No.
S111126 Product Type: Tuning Fork Set
1 options
Catalog No. S111126 Supplier United Scientific™ Supplier No. TFBOX13
Add to Cart
Add to Cart
This tuning fork boxed set of 13 includes a wooden box with foam lining.
  • Each aluminum alloy tuning fork has a specific die-cut place in the foam lining for easy storage.
  • Also contains a mallet specially designed with one hard and one soft end for use with all frequencies of forks.
  • Contains one each of tuning forks with frequencies of 100, 128, 256, 288, 320, 341.3, 384, 426.6, 480, 512, 1024, 2048, and 4096 Hz.
Specifications
Product Type Tuning Fork Set