Learn More
Eisco™ Tuning Fork Set with Case
SCI_ED
Catalog No. p-7230414 Shop All Eisco Products
Change view
Click to view available options
Material:
Aluminum
Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S15127 Aluminum
S15126 Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15127 Supplier Eisco™ Supplier No. PH0744B
Add to Cart
Add to Cart
Tuning Fork Set designed for sound wave lessons and exploration.
  • Set of 13 tuning forks in wooden storage case
  • Tuning forks available in nickled-plated steel or aluminum
  • Plain shanks engraved with frequency and musical note
  • Designed for physics experiments and sound wave studies
  • Gauranteed ± 5% of stated frequency
Includes
  • G# 406, F1 362, B 480, F 341.3, E 320, C1 271, D 288, D# 304.4, C 256, A 426.6, A# 456, G 384, and C 512
Specifications
Product Type Tuning Fork Set
Material Aluminum
For Use With (Application) Sound Wave Lessons and Exploration
Includes 13 Tuning Forks in Various Frequencies, Wooden Storage Case