Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Tuning Forks
SDPSCI_ED

Catalog No. S63122 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Frequency:
256 Hz
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frequency
S63122 256 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S63122 Supplier United Scientific Supplies Supplier No. TFK256/EMD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Frequency and scale letter are stamped on each fork.

  • Made of Aluminum alloy
  • Frequency: 256Hz
  • Scale letter: C'

Specifications

Product Type Tuning Fork
Material Aluminum Alloy
Frequency 256 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.