Learn More
Turbidity 500 NTU Calibration Standard, Formazin Certified Reference Material, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
$102.50 - $159.30
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-100-1728
|
MilliporeSigma Supelco
TURB500100ML |
100 mL |
Each for $102.50
|
|
|||||
11-100-1725
|
MilliporeSigma Supelco
TURB500500ML |
500 mL |
Each for $159.30
|
|
|||||