Learn More
Vernier™ Turbidity Accessories Replacement Kit
SDPSCI_ED
Catalog No. S16271ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Turbidity Sensor
Catalog No. For Use With (Equipment)
S16271ND Turbidity Sensor
1 options
Catalog No. S16271ND Supplier Vernier™ Supplier No. TRBACC
Add to Cart
Add to Cart
Includes
Turbidity Accessories Replacement Kit
Replacement kit of 100 NTU turbidity standard and one empty glass cuvette.
  • Replacement kit
  • 100 NTU turbidity standard
  • One empty glass cuvette
  • For use with Turbidity Sensor
Warranty and Services
This is a consumable and no warranty is included.
Specifications
Type Replacement Kit
Includes 100 NTU turbidity standard and 1 emply glass cuvette
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Turbidity Sensor