Learn More
SPEX CERTIPREP LLC 3P NPW Turbidity, 21mL
Supplier:  SPEX CERTIPREP LLC 3P PEI092
Encompass_Preferred
(NSI by Zeptometrix) A 21 mL concentrate for determination of Turbidity in the NELAC range of 2.0 - 30 NTU. Formazin based. Each container produces 2 liters of sample.
Catalog No. NC9720602