Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Turbidity Sensor
SDPSCI_ED

Catalog No. S16071ND Shop All Vernier Software Products
Change view
Click to view available options
Range:
0 to 200 NTU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S16071ND 0 to 200 NTU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16071ND Supplier Vernier™ Supplier No. TRBBTA
Add to Cart
Add to Cart

Includes

Vernier Turbidity Sensor, 100 NTU turbidity standard, One empty glass cuvette, Material Safety Data Sheet for StablCal Formazin Standard

Requires

Requires an interface and software, sold separately

Use the Turbidity Sensor to measure the turbidity of freshwater or seawater samples and determine water quality. Simple setup and calibration make it easy to use at the collection site or when you return to the classroom.

Range: 0 to 200 NTU Typical Resolution: 0.25 NTU Standard: StablCal Formazin Standard 100 NTU Accuracy: ±2 NTU for readings under 25 NTU ±5% of readings above 25 NTU LED wavelength: 890 nm

Warranty and Services

5 years (no warranty on cuvettes)

Specifications

Includes Sensor, 100 NTU standard, 1 glass cuvette
Grade Middle School, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Accuracy ±2 NTU for readings under 25 NTU, ±5% of readings above 25 NTU
Range 0 to 200 NTU
Resolution 0.25 NTU
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.