Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Turbidity Standard, 100 NTU, Polymer Bead Suspension, Ricca Chemical
SDP

Catalog No. 88061 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Clear PET-G Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
88061 4 L Amber Glass Bottle
88-061-6 500 mL Clear PET-G Bottle
R88060001C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 88061 Supplier Ricca Chemical Company Supplier No. 88061
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9003-53-6 , 26628-22-8 , 7732-18-5
Molecular Formula (C8H8)n
Molecular Weight (g/mol) 104.15
MDL Number MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name ethenylbenzene
SMILES *-CC(-*)C1=CC=CC=C1

Specifications

Packaging Amber Glass Bottle
CAS Min % 98.0, 0.18, 0.01
CAS Max % 100.0, 0.19, 0.01
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 4 L
Chemical Name or Material Turbidity Standard, 100NTU (Nephelometric Turbidity Units) - Polymer Bead Suspension
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.