Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TURBO DNA-free™ Kit

Catalog No. am1907
Encompass_Preferred
Click to view available options
Quantity:
50 Reactions

Includes

TURBO DNase, 10X TURBO DNase Buffer, DNase Inactivation Reagent, Nuclease-free Water

Ideal tool for clearing residual DNA

Kit contains reagents for efficient, complete digestion of DNA along with removal of enzyme and divalent cations post-digestion

  • Hyperactive TURBO DNase is catalytically superior enzyme compared to wild type DNase I
  • Removes trace quantities of DNA that can interfere with RT-PCR
  • Reagent included to completely remove DNase without phenol treatment or heating
  • Method for genomic DNA removal prior to RT-PCR
  • TURBO DNase is recombinant, engineered form of DNase I that is efficient than wild type DNase I in digesting away trace amounts of unwanted DNA
  • TURBO DNase binds DNA substrates 6-fold more tightly than traditional DNase I, making enzyme tool of choice for clearing residual DNA that can generate a false positive signal in RT-PCR applications
  • TURBO DNase now includes an enhancer that increases effectiveness by two orders of magnitude
  • Efficient DNase and divalent cation removal without organic extraction or precipitation
  • Conventional DNase treatment of RNA samples prior to RT-PCR typically call for inactivation of the DNase by phenol:CHCl3 extraction or heating followed by precipitation step to concentrate the RNA
  • Phenol:CHCl3 extractions can be cumbersome and time-consuming
  • Heating the sample to inactivate DNase can lead to chemical degradation of RNA by divalent cations present in DNase buffer
  • TURBO DNA-free Kit circumvents problems using novel DNase Inactivation Reagent
  • For removing the TURBO DNase from reaction, Inactivation Reagent also binds and removes divalent cations from TURBO DNase Reaction Buffer

PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
TURBO DNase
10X TURBO DNase Buffer
DNase Inactivation Reagent
Nuclease-free Water

Store components at -20°C (Nuclease-free water may be stored at room temperature)
Shipping Condition Dry Ice
Enzyme DNase
Compatible Buffer Reaction Buffer
Quantity 50 Reactions
Product Type TURBO DNA Free Kit
Product Line Ambion™, DNA-free
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.