Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TURBO DNase™ (2 U/μL)

Catalog No. am2238
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 U
5000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM2238 1000 U
AM2239 5000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM2238 Supplier Invitrogen™ Supplier No. AM2238
Only null left
Add to Cart
Add to Cart

Efficiently degrades and digests DNA

Cleaves double-stranded DNA nonspecifically to leave 5' phosphorylated oligodeoxynucleotides with an increased affinity for DNA-binding and remains active in presence of salt.

  • Up to 50X more activity and 350% greater catalytic efficiency
  • Efficiently degrades DNA in solutions containing up to 0.25M salt
  • Efficiently digests DNA to oligonucleotides
  • Used in clearing DNA templates from in vitro transcription reactions
  • RNase-free and recombinant in origin
  • Used to clear DNA contamination from RNA samples prior to RT-PCR
  • Conventional DNase I has a poor affinity for DNA and cleaves DNA of low concentration
  • Salt-sensitive; 20 mM NaCl can reduce activity of enzyme by 30%
  • Purified from bovine pancreas, one of richest natural sources of RNase A
  • Threat of contaminating RNase activity in DNase I preparations requires that enzyme be exhaustively purified

TURBO DNA-free (Mfr. No. AM1907) is available. It contains TURBO DNase along with reagents to inactivate the enzyme and remove divalent cations from the sample post-digestion.

PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Content And Storage TURBO™ DNase and 10X Reaction Buffer should be stored at –20°C.
Shipping Condition Dry Ice
Enzyme DNase
Compatible Buffer 10X Reaction Buffer
Quantity 1000 U
Product Type DNase
Product Line Ambion, TURBO
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.