Learn More
Kingfisher Biotech Inc Turkey CCL3 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1526T005
Encompass
The Turkey CCL3 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Turkey CCL3 applications are for cell culture, ELISA standard, and Western Blot Control. Turkey CCL3 yeast-derived recombinant protein can be purchased in multiple sizes. Turkey CCL3 Specifications: (Molecular Weight: 13.5 kDa) (Amino Acid Sequence: IKVAAQQRPS IVQSRALQPP SSSPAHCANV LQTMKGPAAI LTALLLLALC SSAVAHLDEL PTTCCFSYVQ RPIPRNIIKS AYITSSKCRL PAVILVTKKG KEICANPEES WVQNHLELFQ NEEN (124)) (Gene ID: 100544108). For research use only. Made in the USA
Catalog No. 50-253-3199
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.