Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Turkey IgG, H/L Chains Native Protein
SDP

Catalog No. NBP1970331 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1970331 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP1970331 Supplier R&D Systems™ Supplier No. NBP1970331MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

Native Turkey IgG, H/L Chains. The Turkey IgG, H/L Chains Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.

Specifications

Concentration LYOPH
For Use With (Application) ELISA, Immunohistochemistry, Western Blot
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Quantity 1 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Cross Reactivity Turkey IgG, H/L Chains
Species Turkey
Reconstitution Reconstitute with 1.0 ml deionized water.
Product Line Novus
Purity or Quality Grade Immunoelectrophoresis (IEP)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.