Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUSC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577122 Supplier Invitrogen™ Supplier No. PA577122
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

TUSC2 (tumor suppressor candidate 2), also known as FUS1, LGCC or PDAP2, is a 110 amino acid protein that is expressed at high levels in kidney, pancreas, lung, heart and skeletal muscle where it is thought to function as a tumor suppressor. More specifically, TUSC2 inhibits invasive colony formation by inducing G1 cell cycle arrest and ultimately causing apoptosis, thereby preventing tumor growth. The gene encoding TUSC2 maps to human chromosome 3, which houses over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. Key tumor suppressing genes on chromosome 3 include those that encode the apoptosis mediator RASSF1, the cell migration regulator HYAL1 and the angiogenesis suppressor SEMA3B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUSC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TUSC2
Gene Accession No. O75896, Q9WVF8
Gene Alias 1190001E22Rik; AA407686; C3orf11; Fus1; Fus-1 protein; Fusion 1 protein; Lgcc; lung cancer candidiate, FUS; PAP; PDAP2; PDGFA associated protein 2; PDGFA-associated protein 2; Tumor suppressor candidate 2; TUSC 2; Tusc2
Gene Symbols TUSC2
Host Species Rabbit
Immunogen Recombinant full length Human TUSC2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11334, 501052, 80385
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.