Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUSC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551266 Supplier Invitrogen™ Supplier No. PA551266
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total TUSC5 protein.

TUSC5 (tumor suppressor candidate 5), also known as protein located at seventeen-p-thirteen point three 1, LOST1 or IFITMD3 (interferon-induced transmembrane domain-containing protein D3), is a 177 amino acid multi-pass membrane protein that belongs to the CD225 family. Thought to play a role in fat metabolism, TUSC5 is highly expressed in mammary gland, heart, smooth muscle, skeletal muscle and stomach, with lower levels found in lung and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUSC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TUSC5
Gene Accession No. Q8IXB3
Gene Alias BEC-1; brain endothelial cell-derived protein 1; C130069F04Rik; Dispanin subfamily B member 1; DSPB1; IFITMD3; interferon induced transmembrane protein domain containing 3; interferon-induced transmembrane domain-containing protein D3; located at seventeen p thirteen point three 1; Lost1; Protein located at seventeen-p-thirteen point three 1; Trafficking regulator of GLUT4 1; Trarg1; Tumor suppressor candidate 5; tumor suppressor candidate 5 homolog; Tusc5
Gene Symbols TUSC5
Host Species Rabbit
Immunogen Synthetic peptide of human TUSC5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286753
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.