Learn More
Air Boss™ Defense TVI FlexAir™ Accessories for PAPR Systems: Breathing Tube Assembly
Shop All TVI Corporation Products
Specifications
Specifications
| Certifications/Compliance | CE, NIOSH |
| Color | White |
| Material | Tychem QC |
| Product Type | Loose Fitting Visor |
| For Use With (Equipment) | PAPR systems |
| Includes | Drawstring |
| Model | S-40078-W01 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.