Learn More
Specifications
Specifications
| Gene ID (Entrez) | 51330 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56636 |
| Gene Alias | CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12 A, tumor necrosis factor receptor superfamily, member 12 A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 |
| Gene Symbol | TNFRSF12A |
| Quantity | 0.1 mg |
| Research Category | Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
