Learn More
Tween™ 20, Promega
Catalog No. PRW3831 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
2.5 mL
Catalog No. Quantity
PR-W3831 2.5 mL
1 options
Catalog No. PR-W3831 Supplier Promega Corporation Supplier No. W3831
BSA (bovine serum albumin) has been tested and qualified for optimum performance in immunoblotting applications with alkaline phosphatase antibody conjugates.
  • Blot-Qualified BSA and Tween(R) 20 for Use as Blocking Agents for Antibody Detection
  • Blot-Qualified BSA saturates excess sites of antibody binding on membranes for AP antibody conjugates
  • Tween(R) 20 (100%) is used in immunoscreening to dissociate nonspecific interactions with an antibody probe
Specifications
Chemical Name or Material ™Tween™ 20
Quantity 2.5 mL