Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 85

Catalog No. p-4259098
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC334152500 250 mL Glass bottle
AC334150010 1 L Glass bottle
AC334150025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC334152500 Supplier Thermo Scientific Chemicals Supplier No. 334152500
Only null left
Add to Cart

CAS: 9005-70-3 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7 | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-70-3
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
Molecular Weight (g/mol) NaN
MDL Number MFCD01779641
InChI Key ZGEQUSWSYZZNAI-CLFAGFIQNA-N
Synonym Polyoxyethylene (20) Sorbitan Trioleate, Polysorbate 85
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO

Specifications

Color Colorless
Acid Value 2mg KOH/g max.
Physical Form Liquid
Density 1.0200g/mL
Quantity 250 mL
Flash Point >200°C
Hydroxyl Value 38 to 52mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4700 to 1.4720
Saponification Value 83 to 98mg KOH/g
Specific Gravity 1.02
Viscosity 300 mPa.s (25°C)
Water 3% Max.
Solubility Information Solubility in water: soluble. Other solubilities: 100g/L isopropyl alcohol, xylene and mineral oil, insoluble in propylene glycol
Chemical Name or Material Polysorbate 85
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.