Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf twin.tec™ PCR Plate LoBind

Catalog No. 14278232 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric) Well:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Well
14-278-232 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-278-232 Supplier Eppendorf Supplier No. 0030129709
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features a polycarbonate frame and thin-walled polypropylene wells

  • 3-level coding for safe sample and vessel identification
  • Codes on three sides of plates for flexible reading
  • ECC 200 error correction function enables safe code reading with up to 25 % damaged codes
  • Additional, ID-specific information on plates available via Eppendorf DataPort for documentation purposes
  • Direct import into eLabInventory and eLabJournal™ software for convenient handling
  • Import to other database systems possible
  • Manufactured from high-quality polypropylene for highest sample purity
  • Raised well rims enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results
  • Ensures optimal heat transfer and high rigidity
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance ISCC, ACT Label, ISO 9001, ISO 13485, ISO 14001
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Product Line Eppendorf
Color Colorless
Disposable Single-use
Product Type Eppendorf SafeCode Plates
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Conical Bottom (V-well)
No. per Case 25/Cs.
No. per Pack 5/Pk.
Purity or Quality Grade PCR Clean (Non-sterile)
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.