Learn More
Eppendorf™ twin.tec™ PCR Plates BioBased
Catalog No. 01671122 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Clear
Green
Skirt Style:
Skirted
Product Type:
BioBased PCR Plates
No. of Wells:
96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Skirt Style Product Type No. of Wells
01-671-122 Clear Skirted BioBased PCR Plates 96
01-671-121 Green Skirted BioBased PCR Plates 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-671-122 Supplier Eppendorf™ Supplier No. 0030129849
Add to Cart
Add to Cart
Discover the new twin.tec™ PCR Plates BioBased made of bio-based polymer (mass balance approach). These plates allow you to significantly reduce your consumable-related carbon footprint while maintaining identical technical performance.
  • Reduction of consumable-related carbon footprint in the lab
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
  • Available with barcode (upon request)
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Format SBS Standard
Array 8 x 12
Product Line Eppendorf
Color Clear
Product Type BioBased PCR Plates
Skirt Style Skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Quantity 25 Pk.
Fit for Purpose PCR Ready