Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ twin.tec™ Real-time qPCR Plates PROMO

Catalog No. E951022043P Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric) Well:
150 μL
250 μL
Skirt Style:
Semi-skirted
Skirted
Unskirted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Well Skirt Style
E951022043P 250 μL Semi-skirted
E003013270P 150 μL Unskirted
E951022015P 150 μL Skirted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E951022043P Supplier Eppendorf™ Supplier No. 2231001130

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.

  • White wells for better reflection.
  • High mechanical stability.
  • Effective sealing by raised well rims.
  • Skirted and semi-skirted frames.
  • Optimal heat transfer due to reduced wall thickness.
  • Autoclavable (121°C, 20 min.).
  • Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity.
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 25 Plates (5 Bags x 5 Plates)
Color Blue, White Bottom
Product Type PCR Plate
Skirt Style Semi-skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 250 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) RT-PCR Applications
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 25 Pk.
Rimmed Yes
Well Color White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.