Learn More
Eppendorf™ Twin.tec™ Trace PCR Plate 96-well 2-pack PROMO
Catalog No. 01670720PM Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Crystal Blue
Fuchsia
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-670-720PM Fuchsia
01-670-718PM Crystal Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-670-720PM Supplier Eppendorf™ Supplier No. 2231001158
Stay on track - twin.tec™ Trace 96-well PCR plates offer laser-engraved lot IDs, OptiTrack™ grids, and rigid frames for warp-free automation. Stock up with savings on PCR Clean 2-packs!
Eppendorf Twin.tec™ Trace PCR Plate 96 Includes:
50 x Eppendorf twin.tec™ Trace PCR Plate 96, skirted, PCR clean, fuchsia or crystal blue plates
Twin.tec™ Trace PCR Plate 96 Features:
  • Unique laser-engraved optical guiding grid and OptiTrack™ matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 10 Bags x 5 Plates
Product Line Eppendorf
Color Fuchsia
Product Type Trace PCR Plate
Skirt Style Skirted
Material Polypropylene
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Conical Bottom (V-well)
Max. RCF 6000 x g
Quantity 50/Cs.
Well Color Colorless
Fit for Purpose PCR Ready