Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TWIST1 Recombinant Rabbit Monoclonal Antibody (BLR156J)
GREENER_CHOICE

Catalog No. PIMA544497
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544497 100 μL
PIMA544498 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544497 Supplier Invitrogen™ Supplier No. MA544497
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

TWIST1 acts as a transcriptional regulator. It inhibits myogensis by sequestering E proteins, inhibiting trans-activation by MEF2, and inhibiting DNA-binding by MYOD1 through physical interaction. TWIST1 also represses proinflammatory cytokines such as TNFA and IL1B. TWIST also regulates cranial suture patterning and fusion and gene expression differentially depending on dimer composition. Mutations in the gene are implicated in Saethre-Chotzen syndrome, a disease characterized by coronal synostosis, brachycephaly, low front hairline, facial asymmetry, hypertelorism, broad halluces, and clinodactyly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWIST1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR156J
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8-7.4
Gene TWIST1
Gene Accession No. Q15672
Gene Alias AA960487; ACS3; B-HLH DNA binding protein; bHLHa38; BPES2; BPES3; charlie chaplin; class A basic helix-loop-helix protein 38; CRS; CRS1; CSO; H-twist; M-twist; p ACS3; Pde; pdt; Pluridigite; polydactyly EMS; SCS; Ska>m10Jus>; Ska>sup>m10Jus>/sup>; Ska10; Twist; twist basic helix-loop-helix transcription factor 1; twist family bHLH transcription factor 1; twist gene homolog 1; twist homolog 1; TWIST homolog of drosophila; Twist1; twist-related protein 1
Gene Symbols TWIST1
Host Species Rabbit
Immunogen Peptide representing a region between residue 1 and 50 of Human Twist-related protein 1.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7291
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.