Learn More
Bachem b-Defensin 2 (human) Trifluoroacetate
SDP
Supplier:  Bachem 4034693.0500
Encompass_Preferred
b-Defensin 2 (human) Trifluoroacetate, H-Gly-Ile-Gly-Asp-Pro-Val-Thr-Cys-Leu-Lys-Ser-Gly-Ala-Ile-Cys-His-Pro-Val-Phe-Cys-Pro-Arg-Arg-Tyr-Lys-Gln-Ile-Gly-Thr-Cys-Gly-Leu-Pro-Gly-Thr-Lys-Cys-Cys-Lys-Lys-Pro-OH, , C188H305N55O50S6, Mr 4328.23, 0.5mg. b-Defensin 4A (human), Skin-antimicrobial peptide 1 (human). Two b-defensins (hBD1 and hBD2) have been identified in humans. b-defensins are inducible, potent, and selectively killing mainly Gram-negative bacteria and yeast. In addition, b-defensins mobilize cells engaged in adaptive immune responses.
Catalog No. 50-194-6657
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.