Learn More
Diversified Spaces™ Two-Station Steel Workbench with Four Horizontal Locker Openings
SCI_ED
Catalog No. S42435 Shop All Diversified Spaces Products
Change view
Click to view available options
Color:
Black
Gray
Includes:
No vises
One vises
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
S42435 Gray No vises
S42436 Black No vises
S42638 Black One vises
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S42435 Supplier Diversified Spaces™ Supplier No. WBB40V
Add to Cart
Add to Cart
Single-faced heavy gauge steel unit with one section construction and reinforced side panels available with or without vises in gray or black base unit.
  • Base consists of single-faced 20 gauge steel unit
  • Features four horizontal locker openings measuring 53.34D x 45.72W x 38.1cm H (21 x 18 x 15 in.) dimension
  • 18-gauge steel locker doors with air vents, spring hinges, padlock hasp and knockout plug for cylinder lock
  • Maple top with 5.715cm (2.25 in.) thickness
  • Steel lockers with gray baked-enamel finish
  • SEFA compliant and MAS Certified Green
Specifications
Product Type 2-Station Steel Workbench with 4 Horizontal Locker Openings
Warranty Limited Lifetime
Color Gray
Dimensions (D x W x H) 71.12 x 162.56 x 84.45 cm (28 x 64 x 33.25 in.)
Includes No vises
Material Metal