Learn More
Diversified Spaces™ Two-Student Single Storage Cabinet Table
SCI_ED
Catalog No. S38110 Shop All Diversified Spaces Products
Change view
Click to view available options
Includes:
Book Compartment Apron, 2 Locking Drawers
Book Compartment Apron, Plain Base Cabinet
Plain apron, 2 Locking Drawers
Top Style:
ChemGuard
ChemGuard top
Plastic laminate
Solid Epoxy
Solid Phenolic
Weight (English):
275 lb.
326 lb.
335 lb.
350 lb.
355 lb.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Top Style Weight (English)
S38110 Book Compartment Apron, Plain Base Cabinet ChemGuard 275 lb.
S37843 Plain apron, 2 Locking Drawers ChemGuard top 275 lb.
S38109 Book Compartment Apron, Plain Base Cabinet Plastic laminate 275 lb.
S38111 Book Compartment Apron, Plain Base Cabinet Solid Phenolic 326 lb.
S38112 Book Compartment Apron, Plain Base Cabinet Solid Epoxy 350 lb.
S38115 Book Compartment Apron, 2 Locking Drawers ChemGuard 275 lb.
S38116 Book Compartment Apron, 2 Locking Drawers Solid Phenolic 335 lb.
S38117 Book Compartment Apron, 2 Locking Drawers Solid Epoxy 355 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S38110 Supplier Diversified Spaces™ Supplier No. C2102K
Add to Cart
Add to Cart
Allows workspace and storage for two users.
  • Constructed from solid oak and hardwood veneers
  • Features center cabinet with lock and rubber base molding
  • Chemical-resistant, earth-friendly UV finish
Specifications
Product Type 2-Student Single Storage Cabinet Table
Color Oak
Autoclavable Not Autoclavable
No. of Doors 1
No. of Drawers None
No. of Shelves None
With Casters No
Footrest No
With Tilt No
Includes Book Compartment Apron, Plain Base Cabinet
Weight (English) 275 lb.
Dimensions (D x W x H) 24 x 66 x 30 in.
Finish UV
Top Style ChemGuard
Material Red Oak, Hardwood Veneer