Learn More
United Scientific™ Two-Way Table Clamp
SDPSCI_ED
Catalog No. S000851 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Gray
Catalog No. Color
S000851 Gray
1 options
Prop 65
Catalog No. S000851 Supplier United Scientific™ Supplier No. 2WYCLMP
Add to Cart
Add to Cart
Prop 65
Two-way clamp and thumbscrews hold rods firmly in horizontal or vertical positions.
For use with rods up to 3/4 in. in diameter. The cast aluminum, nickel-plated clamp mounts onto tabletops or other surfaces up to 3 in. thick. Screw mechanisms are made of nickel-plated steel. Rods are not included.
  • Hold rods vertically or horizontally
  • Easy to adjust, nickel-plated steel thumbscrews
  • Fits surfaces up to 3 in. thick
Specifications
Type Clamp
Material Cast Aluminum, Nickel-plated Steel
Color Gray
Jaw Type Dual Adjustment