Learn More
Thermo Scientific™ TX-750 Round Bucket Adapters
Description
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Type | Round Bucket Adapter |
| Tube Type | 15 mL Blood Collection Tube |
| Capacity | 84 x 15 mL Blood Collection Tube |
| For Use With (Equipment) | Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube |