Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXNDC11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559467
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559467 Supplier Invitrogen™ Supplier No. PA559467
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSLNHIFIQL ARNLPMDTFT VARIDVSQND LPWEFMVDRL PTVLFFPCNR KDLSVKYPED VPITLPNLLR FILHHSDPAS SPQNVANSPT KECLQ Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.

May act as a redox regulator involved in DUOX proteins folding. The interaction with DUOX1 and DUOX2 suggest that it belongs to a multiprotein complex constituting the thyroid H(2)O(2) generating system. It is however not sufficient to assist DUOX1 and DUOX2 in H(2)O(2) generation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNDC11
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TXNDC11
Gene Accession No. Q6PKC3
Gene Alias 2810408E11Rik; AI427833; EF-hand binding protein 1; EF-hand-binding protein 1; EFP1; RGD1307538; thioredoxin domain containing 11; thioredoxin domain-containing protein 11; Txdc11; Txndc11
Gene Symbols TXNDC11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TXNDC11. Recombinant protein control fragment (Product #RP-98731).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51061
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.