Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ty1 Tag Monoclonal Antibody (BB2)
GREENER_CHOICE

Catalog No. MA523513
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA523513 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA523513 Supplier Invitrogen™ Supplier No. MA523513
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ty1 Tag Monoclonal Antibody for ChIP, Western Blot

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The Ty1 Tag recognizes Ty1-tagged proteins: EVHTNQDPLD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ty1 Tag
Applications ChIP Assay, Western Blot
Classification Monoclonal
Clone BB2
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.4
Gene Alias Ty1 Epitope Tag
Host Species Mouse
Immunogen Ty1 tag (amino acid sequence EVHTNQDPLD).
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.