Description
- Encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
- Integrated socks composed of garment material
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment
- Internal adjustable waist belt system for support and improved fit
- Additional barrier material sewn at knees and over taped for added abrasion resistance when kneeling or crawling
- Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
- Manufactured under specifications that do not contain natural rubber latex
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
- Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Vapor tight zipper
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
- Butyl gloves provide mechanical and physical durability to help protect inner glove
- Gloves are attached with vapor tight ring and clamp system and are replaceable
Specifications
Specifications
| Color | Lime Yellow |
| Material | Tychem |
| Size | 3X-Large |
| Ankle Style | Boot Overflaps |
| Closure Type | Zipper |
| Seam Type | Taped |